2,4,6-Trimethoxyphenylboronic acid ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2,4,6-Trimethoxybenzeneboronic acid, (2,4,6-Trimethoxyphenyl)metaboric acid, Dihydroxy(2,4,6-trimethoxyphenyl)borane
L19837.03
L19837.06
L19837.03EA
2810
CZK
L19837.03
L19837.06
2,4,6-Trimethoxyphenylboronic acid ≥98%
2,4,6-Trimethoxyphenylboronic acid
Purity: 98%
CAS No.: 135159-25-0
Molecular Formula: C9H13BO5
Formula Weight: 212.01
Storage Temperature: -30°C to -10°C
Melting Point: 96-100°
Beilstein Reference: 4315581
MDL No.: MFCD01114642
Vzorec:
C₉H₁₃BO₅ MW: 212,01 g/mol Bod tání: 96…100 °C |
MDL Number:
MFCD01114642 CAS číslo: 135159-25-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...