Tisk…

2,6-Dimethoxyphenylboronic acid ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

B24305.06 B24305.14
B24305.06EA 1710 CZK
B24305.06 B24305.14
2,6-Dimethoxyphenylboronic acid ≥97%
2,6-Dimethoxyphenylboronic acid

Purity: 97+% CAS No.: 23112-96-1 Molecular Formula: C8H11BO4 Formula Weight: 181.98 Melting Point: 106-108° Beilstein Reference: 3032352

Vzorec: C₈H₁₁BO₄
Bod tání: 106…108 °C
Storage Temperature: Ambient
MDL Number: MFCD01318987
CAS číslo: 23112-96-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR