3-Methoxyphenylboronic acid ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
(3-Methoxyphenyl)metaboric acid, Dihydroxy(3-methoxyphenyl)borane, 3-Methoxybenzeneboronic acid
B24412.03
B24412.06
B24412.14
AB134067.0001
B24412.03EA
1530
CZK
B24412.03
B24412.06
B24412.14
ABCRAB134067.0001
3-Methoxyphenylboronic acid ≥97%
3-Methoxyphenylboronic acid
Purity: 97%
CAS No.: 10365-98-7
Molecular Formula: C7H9BO3
Formula Weight: 151.96
Melting Point: 158-162°
Beilstein Reference: 2831223
Vzorec:
C₇H₉BO₃ MW: 151,96 g/mol Bod tání: 158…162 °C Storage Temperature: Ambient |
MDL Number:
MFCD00161359 CAS číslo: 10365-98-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...