Tisk…

2-Methoxyphenylboronic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-Methoxybenzeneboronic acid, (2-Methoxyphenyl)metaboric acid, Dihydroxy(2-methoxyphenyl)borane

M-Clarity™ quality level = MQ100

445231-1G 445231-5G
SIAL445231-1GEA 938 CZK
SIAL445231-1G SIAL445231-5G
2-Methoxyphenylboronic acid, Sigma-Aldrich®
2-Methoxyphenylboronic acid
Vzorec: C₇H₉BO₃
MW: 151.96
Bod tání: 165
MDL Number: MFCD00236047
CAS číslo: 5720-06-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR