2-Methoxyphenylboronic acid ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
2-Methoxybenzeneboronic acid, (2-Methoxyphenyl)metaboric acid, Dihydroxy(2-methoxyphenyl)borane
B21071.03
B21071.06
B21071.14
B21071.03EA
862
CZK
B21071.03
B21071.06
B21071.14
2-Methoxyphenylboronic acid ≥97%
2-Methoxyphenylboronic acid
Purity: 97%
CAS No.: 5720-06-9
Molecular Formula: C7H9BO3
Formula Weight: 151.96
Melting Point: 102-106°
Beilstein Reference: 3030981
Vzorec:
C₇H₉BO₃ Bod tání: 102…106 °C Storage Temperature: Ambient |
MDL Number:
MFCD00236047 CAS číslo: 5720-06-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...