Tisk…

3-Nitrophenylboronic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (3-Nitrophenyl)metaboric acid, Dihydroxy(3-nitrophenyl)borane, 3-Nitrobenzeneboronic acid

M-Clarity™ quality level = MQ100

325104-1G 325104-5G
SIAL325104-1GEA 748 CZK
SIAL325104-1G SIAL325104-5G
3-Nitrophenylboronic acid, Sigma-Aldrich®
3-Nitrophenylboronic acid
Vzorec: C₆H₆BNO₄
MW: 166,93 g/mol
Bod tání: 290…291 °C
MDL Number: MFCD00007193
CAS číslo: 13331-27-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR