Tisk…

2-Nitrophenylboronic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Dihydroxy(2-nitrophenyl)borane, (2-Nitrophenyl)boronic acid, 2-Nitrobenzeneboronic acid, (2-Nitrophenyl)metaboric acid

M-Clarity™ quality level = MQ100

673862-1G 673862-5G
SIAL673862-1GEA 0 CZK
SIAL673862-1G SIAL673862-5G
2-Nitrophenylboronic acid, Sigma-Aldrich®
2-Nitrophenylboronic acid
Vzorec: C₆H₆BNO₄
MW: 166,93 g/mol
MDL Number: MFCD00161358
CAS číslo: 5570-19-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR