2-Nitrophenylboronic acid 96%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Dihydroxy(2-nitrophenyl)borane, (2-Nitrophenyl)boronic acid, 2-Nitrobenzeneboronic acid, (2-Nitrophenyl)metaboric acid
L17988.06
L17988.03
L17988.06EA
11290
CZK
L17988.06
L17988.03
2-Nitrophenylboronic acid 96%
2-Nitrophenylboronic acid
Purity: 96%
CAS No.: 5570-19-4
Molecular Formula: C6H6BNO4
Formula Weight: 166.93
Melting Point: 149-151°
Beilstein Reference: 3032097
Vzorec:
C₆H₆BNO₄ MW: 166.93 Bod tání: 149…151 °C Storage Temperature: Ambient |
MDL Number:
MFCD00161358 CAS číslo: 5570-19-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...