Tisk…

3-Cyanophenylboronic acid, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: (3-Cyanophenyl)metaboric acid, Dihydroxy(3-cyanophenyl)borane, 3-Cyanobenzeneboronic acid

M-Clarity™ quality level = MQ100

513016-1G 513016-5G
SIAL513016-1GEA 4440 CZK
SIAL513016-1G SIAL513016-5G
3-Cyanophenylboronic acid, Sigma-Aldrich®
3-Cyanophenylboronic acid
Vzorec: C₇H₆BNO₂
MW: 146,94 g/mol
MDL Number: MFCD01318967
CAS číslo: 150255-96-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR