Tisk…

Phenylboronic acid, Sigma-Aldrich®

Dodavatel: Merck
Varování


M-Clarity™ quality level = MQ200

P20009-10G P20009-250G P20009-50G
SIALP20009-10GEA 1830 CZK
SIALP20009-10G SIALP20009-250G SIALP20009-50G
Phenylboronic acid, Sigma-Aldrich®
Phenylboronic acid
Vzorec: C₆H₇BO₂
Bod tání: 217…220 °C
Storage Temperature: Ambient
MDL Number: MFCD00002103
CAS číslo: 98-80-6
EINECS: 202-701-9
Merck Index: 13,01068

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR