Tisk…

Phenylboronic acid, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: Phenylmetaboric acid, Dihydroxy(phenyl)borane, Phenyl boric acid, Benzeneboronic acid

M-Clarity™ quality level = MQ200

78181-10G 78181-1G 78181-50G
SIAL78181-10GEA 1940 CZK
SIAL78181-10G SIAL78181-1G SIAL78181-50G
Phenylboronic acid, Sigma-Aldrich®
Phenylboronic acid
Vzorec: C₆H₇BO₂
MW: 121,93 g/mol
Bod tání: 217…220 °C
Storage Temperature: Ambient
MDL Number: MFCD00002103
CAS číslo: 98-80-6
EINECS: 202-701-9
Merck Index: 13,01068

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR