Phenylboronic acid ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Phenylmetaboric acid, Dihydroxy(phenyl)borane, Phenyl boric acid, Benzeneboronic acid
A14257.30
A14257.09
A14257.18
A14257.30EA
22690
CZK
A14257.30
A14257.09
A14257.18
Phenylboronic acid ≥98%
Phenylboronic acid
Purity: 98+%
CAS No.: 98-80-6
Molecular Formula: C6H7BO2
Formula Weight: 121.93
Melting Point: 214-219°
Beilstein Reference: 970972
Merck Reference: 14,1068
MDL No.: MFCD00002103
Vzorec:
C₆H₇BO₂ Bod tání: 214…219 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002103 CAS číslo: 98-80-6 EINECS: 202-701-9 Merck Index: 14,1068 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...