Tisk…

Piperonylic acid, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

P49805-25G P49805-5G
SIALP49805-25GEA 2620 CZK
SIALP49805-25G SIALP49805-5G
Piperonylic acid, Sigma-Aldrich®
Piperonylic acid
Vzorec: C₈H₆O₄
Bod varu: 180 °C (1 mmHg)
Bod tání: 232…235 °C
MDL Number: MFCD00005830
CAS číslo: 94-53-1
EINECS: 202-342-8
Merck Index: 12,07630

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR