Tisk…

Piperonylic acid ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 3,4-(Methylenedioxy)benzoic acid, Benzo[d][1,3]dioxole-5-carboxylic acid, 1,3-Benzodioxole-5-carboxylic acid

A14105.14 A14105.22 A14105.36
A14105.14EA 2320 CZK
A14105.14 A14105.22 A14105.36
Piperonylic acid ≥98%
Piperonylic acid

Purity: 98+% CAS No.: 94-53-1 Molecular Formula: C8H6O4 Formula Weight: 166.13 Melting Point: 230-235° Beilstein Reference: 150206 Merck Reference: 14,7477 MDL No.: MFCD00005830

Vzorec: C₈H₆O₄
MW: 166,13 g/mol
Bod varu: 180 °C (1 mmHg)
Bod tání: 230…235 °C
Storage Temperature: Ambient
MDL Number: MFCD00005830
CAS číslo: 94-53-1
EINECS: 202-342-8
Merck Index: 14,7477

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR