Tisk…

Dichlone ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 2,3-Dichloro-1,4-naphthoquinone

A13378.14 A13378.22 A13378.36
A13378.14EA 718 CZK
A13378.14 A13378.22 A13378.36
Dichlone ≥98%
Dichlone

Purity: 98% CAS No.: 117-80-6 Molecular Formula: C10H4Cl2O2 Formula Weight: 227.05 Melting Point: 193-196° subl. Boiling Point: 275°/2mm Merck Reference: 14,3045 UN No.: 2811 MDL No.: MFCD00001677

Vzorec: C₁₀H₄Cl₂O₂
MW: 227.05
Bod varu: 275 °C (2 mmHg)
Bod tání: 193…196 °C (subl.)
Hustota: 1.44 g/cm³ (16 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001677
CAS číslo: 117-80-6
EINECS: 204-210-5
UN: 2811
ADR: 6.1 III
Merck Index: 13,03071

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR