1,2,3,4-Tetrahydronaftalen ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
A14962.0B
A14962.0I
A14962.30
A14962.0BEA
1930
CZK
A14962.0B
A14962.0I
A14962.30
1,2,3,4-Tetrahydronaftalen ≥97%
1,2,3,4-Tetrahydronaftalen
Purity: 97%
CAS No.: 119-64-2
Molecular Formula: C10H12
Formula Weight: 132.21
Flash Point: 77°(171°F)
Refractive Index: 1.5400
Melting Point: -36°
Boiling Point: 206-207°
Beilstein Reference: 1446407
Merck Reference: 14,9221
UN No.: 3082
MDL No.: MFCD00001733
Vzorec:
C₁₀H₁₂ MW: 132,21 g/mol Bod varu: 206…207 °C Bod tání: –36 °C Hustota: 0,966 Bod vzplanutí: 77 °C (171 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001733 CAS číslo: 119-64-2 EINECS: 204-340-2 UN: 3082 ADR: 9,III Merck Index: 14,9221 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...