1-Methylisatin ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
A12942.03
A12942.06
A12942.14
A12942.03EA
831
CZK
A12942.03
A12942.06
A12942.14
1-Methylisatin ≥97%
1-Methylisatin
Purity: 97%
CAS No.: 2058-74-4
Molecular Formula: C9H7NO2
Formula Weight: 161.16
Melting Point: 129-133°
Beilstein Reference: 128280
Vzorec:
C₉H₇NO₂ Bod tání: 129…133 °C Hustota: 1,4 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00005812 CAS číslo: 2058-74-4 EINECS: 218-164-9 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...