5-Chloro-7-methylisatin ≥97%
Dodavatel: Thermo Scientific
L11682.03
L11682.06
L11682.MD
L11682.03EA
3230
CZK
L11682.03
L11682.06
L11682.MD
5-Chloro-7-methylisatin ≥97%
5-Chloro-7-methylisatin
Purity: 97%
CAS No.: 14389-06-1
Molecular Formula: C9H6ClNO2
Formula Weight: 195.61
Melting Point: ca 283° dec.
Beilstein Reference: 153061
Vzorec:
C₉H₆ClNO₂ Bod tání: ∼ 283 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00047218 CAS číslo: 14389-06-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...