Indoline ≥99%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2,3-Dihydroindol
A11000.0B
A11000.18
A11000.30
A11000.PG
A11000.0BEA
20350
CZK
A11000.0B
A11000.18
A11000.30
A11000.PG
Indoline ≥99%
Indoline
Purity: 99%
CAS No.: 496-15-1
Molecular Formula: C8H9N
Formula Weight: 119.17
Flash Point: 92°(197°F)
Refractive Index: 1.5930
Odour: Odourless
Melting Point: -21°
Boiling Point: 220-221°
Beilstein Reference: 111915
MDL No.: MFCD00005705
Vzorec:
C₈H₉N MW: 119,17 g/mol Bod varu: 220…221 °C Bod tání: –21 °C Hustota: 1,062 Bod vzplanutí: 92 °C (197 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005705 CAS číslo: 496-15-1 EINECS: 207-816-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...