Tisk…

5-Iodoisatin, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 5-Iodoindoline-2,3-dione, 5-Iodo-1H-indole-2,3-dione

M-Clarity™ quality level = MQ100

140589-25G 140589-5G
SIAL140589-25GEA 5800 CZK
SIAL140589-25G SIAL140589-5G
5-Iodoisatin, Sigma-Aldrich®
5-Iodoisatin
Vzorec: C₈H₄INO₂
MW: 273.03
MDL Number: MFCD00016899
CAS číslo: 20780-76-1
EINECS: 244-035-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR