Tisk…

7-Fluoroisatin ≥97%

Dodavatel: Thermo Scientific
Varování

Synonyms: 7-Fluoro-1H-indole-2,3-dione, 7-Fluoroindoline-2,3-dione

H26827.03 H26827.06 H26827.14
H26827.03EA 972 CZK
H26827.03 H26827.06 H26827.14
7-Fluoroisatin ≥97%
7-Fluoroisatin

Purity: 97% CAS No.: 317-20-4 Molecular Formula: C8H4FNO2 Formula Weight: 165.12 Melting Point: 193-195° MDL No.: MFCD01569508

Vzorec: C₈H₄FNO₂
MW: 165,12 g/mol
Bod tání: 193…195 °C
Storage Temperature: Ambient
MDL Number: MFCD01569508
CAS číslo: 317-20-4
EINECS: None

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR