Izatin, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2,3-Indolinedione, Indoline-2,3-dione, 1H-Indole-2,3-dione
M-Clarity™ quality level = MQ100
114618-100G
114618-500G
114618-5G
SIAL114618-100GEA
1230
CZK
SIAL114618-100G
SIAL114618-500G
SIAL114618-5G
Izatin, Sigma-Aldrich®
Izatin
Vzorec:
C₈H₅NO₂ MW: 147,13 g/mol Bod varu: 342 °C (1013 hPa) Bod tání: 198…202 °C Hustota: 1,472 g/cm³ (20 °C) Bod vzplanutí: 220 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005718 CAS číslo: 91-56-5 EINECS: 202-077-8 Merck Index: 13,05119 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...