Tisk…

Izatin ≥98%

Dodavatel: Thermo Scientific

Synonyms: 2,3-Indolinedione, Indoline-2,3-dione, 1H-Indole-2,3-dione

A12468.0E A12468.22 A12468.36
A12468.0EEA 15550 CZK
A12468.0E A12468.22 A12468.36
Izatin ≥98%
Izatin

Purity: 98% CAS No.: 91-56-5 Molecular Formula: C8H5NO2 Formula Weight: 147.13 Flash Point: 220°(428°F) Odour: Odourless pH: 7 Melting Point: ca 199-200° Beilstein Reference: 383659 Merck Reference: 14,5104 MDL No.: MFCD00005718

Vzorec: C₈H₅NO₂
MW: 147,13 g/mol
Bod varu: 342 °C (1013 hPa)
Bod tání: ∼199…200 °C
Hustota: 1,472 g/cm³ (20 °C)
Bod vzplanutí: 220 °C (428 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005718
CAS číslo: 91-56-5
EINECS: 202-077-8
Merck Index: 14,5104

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR