Tisk…

5-Bromoisatin, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 5-Bromoindoline-2,3-dione, 5-Bromo-1H-indole-2,3-dione

M-Clarity™ quality level = MQ100

476994-25G 476994-5G
SIAL476994-25GEA 3540 CZK
SIAL476994-25G SIAL476994-5G
5-Bromoisatin, Sigma-Aldrich®
5-Bromoisatin
Vzorec: C₈H₄BrNO₂
MW: 226,03 g/mol
MDL Number: MFCD00149345
CAS číslo: 87-48-9
EINECS: 201-747-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR