5-Bromoisatin ≥90%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
5-Bromoindoline-2,3-dione, 5-Bromo-1H-indole-2,3-dione
A13641.18
A13641.22
A13641.36
A13641.18EA
2760
CZK
A13641.18
A13641.22
A13641.36
5-Bromoisatin ≥90%
5-Bromoisatin
Purity: 90+%
CAS No.: 87-48-9
Molecular Formula: C8H4BrNO2
Formula Weight: 226.03
Melting Point: ca 250° dec.
Beilstein Reference: 383760
Vzorec:
C₈H₄BrNO₂ MW: 226,03 g/mol Bod tání: ∼ 250 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00149345 CAS číslo: 87-48-9 EINECS: 201-747-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...