Benzimidazole, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
1H-Benzo[d]imidazole, 1H-Benzimidazole
M-Clarity™ quality level = MQ200
194123-100G
194123-500G
194123-5G
SIAL194123-100GEA
1810
CZK
SIAL194123-100G
SIAL194123-500G
SIAL194123-5G
Benzimidazole, Sigma-Aldrich®
Benzimidazole
Vzorec:
C₇H₆N₂ MW: 118,14 g/mol Bod varu: 360 °C (1013 hPa) Bod tání: 170…173 °C Hustota: 1,23 g/cm³ (20 °C) |
MDL Number:
MFCD00005585 CAS číslo: 51-17-2 EINECS: 200-081-4 Merck Index: 13,01082 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...