Morfolin 99,0 ACS
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
31984.36
31984.57
31984.36EA
1950
CZK
31984.36
31984.57
Morfolin 99,0 ACS
Morfolin
Purity: 99.0% min
CAS No.: 110-91-8
Molecular Formula: C4H9NO
Formula Weight: 87.12
Flash Point: 35°(95°F)
Physical Form: Liquid
Density: 1.007
Refractive Index: 1.4540
Sensitivity: Hygroscopic
Appearance: Colourless
Odour: Amine-like
Melting Point: -5°
Boiling Point: 129°
Beilstein Reference: 102549
Merck Reference: 14,6277
UN No.: 2054
MDL No.: MFCD00005972
Vzorec:
C₄H₉NO MW: 87.12 Bod varu: 129 °C Bod tání: –5 °C Hustota: 1.007 Bod vzplanutí: 35 °C (95 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005972 CAS číslo: 110-91-8 EINECS: 203-815-1 UN: 2054 ADR: 8 I Merck Index: 14,6277 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...