1-Phenylpyrrolidine ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
N-Phenylpyrrolidine
L05239.06
L05239.14
L05239.06EA
2380
CZK
L05239.06
L05239.14
1-Phenylpyrrolidine ≥98%
1-Phenylpyrrolidine
Purity: 98+%
CAS No.: 4096-21-3
Molecular Formula: C10H13N
Formula Weight: 147.22
Refractive Index: 1.5820
Boiling Point: 133-134°/19mm
MDL No.: MFCD00015897
Vzorec:
C₁₀H₁₃N MW: 147,22 g/mol Bod varu: 133…134 °C (19 mmHg) Bod tání: 11 °C Hustota: 1,018 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00015897 CAS číslo: 4096-21-3 EINECS: 223-849-0 UN: 3287 ADR: 6.1,III |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...