4-Methylpiperidine, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
M73206-100ML
M73206-500ML
M73206-5ML
SIALM73206-100MLEA
1220
CZK
SIALM73206-100ML
SIALM73206-500ML
SIALM73206-5ML
4-Methylpiperidine, Sigma-Aldrich®
4-Methylpiperidine
Vzorec:
C₆H₁₃N Bod varu: 124 °C (1013 hPa) Bod tání: –4 °C Hustota: 0,84 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006005 CAS číslo: 626-58-4 EINECS: 210-954-1 UN: 2733 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...