4-Methylpiperidine ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Purity: 98+%
CAS No.: 626-58-4
Molecular Formula: C6H13N
Formula Weight: 99.18
Flash Point: 13°(55°F)
Refractive Index: 1.4458
Odour: Amine-like
pH: 13
Melting Point: 4-5°
Boiling Point: 123-124°
Beilstein Reference: 1083
UN No.: 2733
MDL No.: MFCD00006005
Vzorec:
C₆H₁₃N Bod varu: 123…124 °C Bod tání: 4…5 °C Hustota: 0,838 Bod vzplanutí: 13 °C (55 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006005 CAS číslo: 626-58-4 EINECS: 210-954-1 UN: 2733 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...