3-Methylpiperidine, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
M73001-100G
M73001-5G
SIALM73001-100GEA
2460
CZK
SIALM73001-100G
SIALM73001-5G
3-Methylpiperidine, Sigma-Aldrich®
3-Methylpiperidine
Vzorec:
C₆H₁₃N MW: 99,18 g/mol Bod varu: 125 °C (1013 hPa) Bod tání: –24 °C Hustota: 0,8635 g/cm³ (0 °C) |
MDL Number:
MFCD00005994 CAS číslo: 626-56-2 EINECS: 210-953-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...