1-methyl-2-pyrolidon 99,5, Biograde DNA_synthesis_peptides
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
1-Methyl-2-pyrrolidinone, N-Methyl-2-pyrrolidinone,NMP, 1-Methyl-2-pyrrolidone, NMP, Methyl Pyrrolidone
Argon_filled
Purity: 99.5%
CAS No.: 872-50-4
Molecular Formula: C5H9NO
Formula Weight: 99.13
Flash Point: 86°(187°F)
Physical Form: Liquid, packaged under Argon
Density: 1.033
Refractive Index: 1.4705
Sensitivity: Light Sensitive and Hygroscopic
Appearance: Colourless to pale yellow
Odour: Amine-like
Melting Point: -24°
Boiling Point: 204°
Beilstein Reference: 106420
Merck Reference: 14,6117
MDL No.: MFCD00003193
Suitable for Biotech applications
Vzorec:
C₅H₉NO MW: 99.13 Bod varu: 204 °C Bod tání: –24 °C Hustota: 1.033 Bod vzplanutí: 86 °C (187 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003193 CAS číslo: 872-50-4 EINECS: 212-828-1 Merck Index: 14,6117 |