Piperidin, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
Hexahydropyridine, Hexazane, Pentamethyleneimine
M-Clarity™ quality level = MQ200
411027-100ML
411027-1L
SIAL411027-100MLEA
2260
CZK
SIAL411027-100ML
SIAL411027-1L
Piperidin, Sigma-Aldrich®
Piperidin
Vzorec:
C₅H₁₁N MW: 85,15 g/mol Bod varu: 107,3 °C (1013 hPa) Bod tání: –10 °C Hustota: 0,8622 g/cm³ (20 °C) Bod vzplanutí: 16 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005979 CAS číslo: 110-89-4 EINECS: 203-813-0 UN: 2401 ADR: 8,I Merck Index: 12,07621 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...