Tisk…

N-Iodosuccinimide ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 1-Iodopyrrolidine-2,5-dione

A14320.06 A14320.22 A14320.14 AB109218.0100
A14320.06EA 1650 CZK
A14320.06 A14320.22 A14320.14 ABCRAB109218.0100
N-Iodosuccinimide ≥97%
N-Iodosuccinimide

Purity: 97% CAS No.: 516-12-1 Molecular Formula: C4H4INO2 Formula Weight: 224.99 Storage Temperature: 2°C to 8°C Physical Form: Powder Density: 2.245 Sensitivity: Moisture and Light Sensitive Appearance: White to pale brown Melting Point: ca 196° dec. Beilstein Reference: 113917 Merck Reference: 14,5045 MDL No.: MFCD00005512

Vzorec: C₄H₄INO₂
Bod varu: 356 °C (1013 hPa)
Hustota: 2,245
Storage Temperature: Refrigerator
MDL Number: MFCD00005512
CAS číslo: 516-12-1
EINECS: 208-221-6
Merck Index: 14,5045

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR