Tisk…

N-Hydroxysuccinimide, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 1-Hydroxypyrrolidine-2,5-dione, NHS, HOSu

M-Clarity™ quality level = MQ200

56480-100G 56480-1KG 56480-25G
SIAL56480-100GEA 4630 CZK
SIAL56480-100G SIAL56480-1KG SIAL56480-25G
N-Hydroxysuccinimide, Sigma-Aldrich®
N-Hydroxysuccinimide
Vzorec: C₄H₅NO₃
Bod varu: 363 °C (1013 hPa)
Bod tání: 93…95 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005516
CAS číslo: 6066-82-6
EINECS: 228-001-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR