Tisk…

N-Bromsukcinimid ≥99%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: NBS, 1-Bromopyrrolidine-2,5-dione

A15922.0B A15922.0I A15922.30
A15922.0BEA 4430 CZK
A15922.0B A15922.0I A15922.30
N-Bromsukcinimid ≥99%
N-Bromsukcinimid

Purity: 99% CAS No.: 128-08-5 Molecular Formula: C4H4BrNO2 Formula Weight: 177.99 Odour: Pungent Melting Point: 175-180° dec. Beilstein Reference: 113916 Merck Reference: 14,1438 UN No.: 3261 MDL No.: MFCD00005510


Allylic bromination reagent. Cleaves peptides at tryptophyl residues

Vzorec: C₄H₄BrNO₂
MW: 177,99 g/mol
Bod varu: 339 °C (1013 hPa)
Bod tání: 175…180 °C (rozkl.)
Hustota: 2,098
Storage Temperature: Ambient
MDL Number: MFCD00005510
CAS číslo: 128-08-5
EINECS: 204-877-2
UN: 3261
ADR: 8,III
Merck Index: 14,1438

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR