9,10-Diphenylanthracene, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
D205001-1G-A
D205001-5G-A
SIALD205001-1G-AEA
4020
CZK
SIALD205001-1G-A
SIALD205001-5G-A
9,10-Diphenylanthracene, Sigma-Aldrich®
9,10-Diphenylanthracene
Vzorec:
C₂₆H₁₈ MW: 330,43 g/mol Bod varu: 485 °C (1013 hPa) Bod tání: 245 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001253 CAS číslo: 1499-10-1 EINECS: 216-105-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...