9-Vinylanthracene, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
V1708-1G
V1708-5G
SIALV1708-1GEA
5200
CZK
SIALV1708-1G
SIALV1708-5G
9-Vinylanthracene, Sigma-Aldrich®
9-Vinylanthracene
Vzorec:
C₁₆H₁₂ MW: 204,27 g/mol Bod varu: 377 °C (1013 hPa) Bod tání: 64 °C Hustota: 1,269 g/cm³ (–150.15 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00003576 CAS číslo: 2444-68-0 EINECS: 219-486-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...