Tisk…

1-Aminoanthracene, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: 1-Anthramine, Anthracen-1-amine

M-Clarity™ quality level = MQ100

A38606-1G A38606-5G
SIALA38606-1GEA 3540 CZK
SIALA38606-1G SIALA38606-5G
1-Aminoanthracene, Sigma-Aldrich®
1-Aminoanthracene
Vzorec: C₁₄H₁₁N
MW: 193,25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003579
CAS číslo: 610-49-1
EINECS: 210-225-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR