5-Methyl-1-hexyne, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
662860-25G
662860-5G
SIAL662860-25GEA
6410
CZK
SIAL662860-25G
SIAL662860-5G
5-Methyl-1-hexyne, Sigma-Aldrich®
5-Methyl-1-hexyne
Vzorec:
C₇H₁₂ MW: 96,17 g/mol Bod varu: 92 °C (1013 hPa) Bod tání: –125 °C Hustota: 0,7274 g/cm³ (20 °C) |
MDL Number:
MFCD00041610 CAS číslo: 2203-80-7 UN: 1993 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...