Tisk…

4-Methyl-1-pentyne ≥97%

Dodavatel: Thermo Scientific
Nebezpečí

L05537.03 L05537.06
L05537.03EA 1680 CZK
L05537.03 L05537.06
4-Methyl-1-pentyne ≥97%
4-Methyl-1-pentyne

Purity: 97% CAS No.: 7154-75-8 Molecular Formula: (CH3)2CHCH2C≡CH Formula Weight: 82.15 Flash Point: -24°(-11°F) Refractive Index: 1.3930 Boiling Point: 61-62° Beilstein Reference: 1731077 UN No.: 3295 MDL No.: MFCD00039857

Vzorec: C₆H₁₀
MW: 82,15 g/mol
Bod varu: 61…62 °C
Bod tání: –110,3 °C
Hustota: 0,698
Bod vzplanutí: –24 °C (–11 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039857
CAS číslo: 7154-75-8
UN: 3295
ADR: 3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR