Tisk…

2-Methyl-3-butyne-2-ol ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

L07593.AE L07593.AP L07593.0F AB172513.0100
L07593.AEEA 872 CZK
L07593.AE L07593.AP L07593.0F ABCRAB172513.0100
2-Methyl-3-butyne-2-ol ≥98%
2-Methyl-3-butyne-2-ol

Purity: 98% CAS No.: 115-19-5 Molecular Formula: C5H8O Formula Weight: 84.12 Flash Point: 22°(72°F) Refractive Index: 1.4210 Melting Point: 2-3° Boiling Point: 103-104° Beilstein Reference: 635746 Merck Reference: 14,6034 UN No.: 1987 MDL No.: MFCD00004467

Vzorec: C₅H₈O
Bod varu: 103…104 °C
Bod tání: 2…3 °C
Hustota: 0,863
Bod vzplanutí: 22 °C (72 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004467
CAS číslo: 115-19-5
EINECS: 204-070-5
UN: 1987
ADR: 3,II
Merck Index: 14,6034

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR