3-Butyne-1-ol ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
4-Hydroxy-1-butyne, But-3-yn-1-ol, 3-Butyn-1-ol, 1-Hydroxy-3-butyne
A11477.06
A11477.14
A11477.30
A11477.06EA
1380
CZK
A11477.06
A11477.14
A11477.30
3-Butyne-1-ol ≥98%
3-Butyne-1-ol
Purity: 98+%
CAS No.: 927-74-2
Molecular Formula: C4H6O
Formula Weight: 70.09
Flash Point: 36°(96°F)
Refractive Index: 1.4405
Odour: Characteristic
Melting Point: -63°
Boiling Point: 129-130°
Beilstein Reference: 773710
UN No.: 1987
MDL No.: MFCD00002955
Vzorec:
C₄H₆O Bod varu: 129…130 °C Bod tání: –63 °C Hustota: 0,927 Bod vzplanutí: 36 °C (96 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002955 CAS číslo: 927-74-2 EINECS: 213-161-9 UN: 1987 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...