Propargylalkohol ≥99%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
2-Propyn-1-ol, Prop-2-yn-1-ol
A10295.0F
A10295.AE
A10295.AP
A10295.0FEA
6760
CZK
A10295.0F
A10295.AE
A10295.AP
Propargylalkohol ≥99%
Propargylalkohol
Purity: 99%
CAS No.: 107-19-7
Molecular Formula: C3H4O
Formula Weight: 56.06
Storage Temperature: 2°C to 8°C
Flash Point: 33°(91°F)
Refractive Index: 1.4320
Melting Point: -53°
Boiling Point: 114-115°
Beilstein Reference: 506003
Merck Reference: 14,7809
UN No.: 2929
MDL No.: MFCD00002912
Vzorec:
C₃H₄O MW: 56,06 g/mol Bod varu: 114…115 °C Bod tání: –53 °C Hustota: 0,949 Bod vzplanutí: 33 °C (91 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00002912 CAS číslo: 107-19-7 EINECS: 203-471-2 UN: 2929 ADR: 6.1,II Merck Index: 14,7809 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...