Tisk…

5-Hexynoic acid 96%

Dodavatel: Thermo Scientific
Nebezpečí

B25132.03 B25132.06 B25132.14
B25132.03EA 1390 CZK
B25132.03 B25132.06 B25132.14
5-Hexynoic acid 96%
5-Hexynoic acid

Purity: 97% CAS No.: 53293-00-8 Molecular Formula: HC≡C(CH2)3CO2H Formula Weight: 112.13 Flash Point: 110°(230°F) Refractive Index: 1.4495 Boiling Point: 73-74°/0.2mm Beilstein Reference: 1743192 UN No.: 3265 MDL No.: MFCD00066346

Vzorec: C₆H₈O₂
MW: 112,13 g/mol
Bod varu: 73…74 °C (0,2 mmHg)
Bod tání: 41 °C
Hustota: 0,990
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00066346
CAS číslo: 53293-00-8
UN: 3265
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR