Tisk…

2-Octynoic acid ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Oct-2-ynoic acid

B21957.06 B21957.14 B21957.22
B21957.06EA 3000 CZK
B21957.06 B21957.14 B21957.22
2-Octynoic acid ≥98%
2-Octynoic acid

Purity: 98% CAS No.: 5663-96-7 Molecular Formula: CH3(CH2)4C≡CCO2H Formula Weight: 140.18 Flash Point: 101°(214°F) Refractive Index: 1.4600 Boiling Point: 148-149°/19mm UN No.: 3265 MDL No.: MFCD00004364

Vzorec: C₈H₁₂O₂
MW: 140,18 g/mol
Bod varu: 148…149 °C (19 mmHg)
Bod tání: 5 °C
Hustota: 0,961
Bod vzplanutí: 101 °C (214 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004364
CAS číslo: 5663-96-7
EINECS: 227-121-3
UN: 3265
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR