Tisk…

17-Octadecynoic acid, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

O8382-25MG O8382-5MG
SIALO8382-25MGEA 41670 CZK
SIALO8382-25MG SIALO8382-5MG
17-Octadecynoic acid, Sigma-Aldrich®
17-Octadecynoic acid
Vzorec: C₁₈H₃₂O₂
MW: 280,45 g/mol
CAS číslo: 34450-18-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR