Tisk…

3-Butyne-2-one ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

L05527.03 L05527.06 L05527.14
L05527.03EA 2120 CZK
L05527.03 L05527.06 L05527.14
3-Butyne-2-one ≥98%
3-Butyne-2-one

Purity: 98% CAS No.: 1423-60-5 Molecular Formula: C4H4O Formula Weight: 68.08 Storage Temperature: 2°C to 8°C Flash Point: -1°(30°F) Refractive Index: 1.4060 Boiling Point: 84-86° Beilstein Reference: 605353 UN No.: 1992 MDL No.: MFCD00008775

Vzorec: C₄H₄O
Bod varu: 84…86 °C
Hustota: 0,870
Bod vzplanutí: –1 °C (30 °F)
MDL Number: MFCD00008775
CAS číslo: 1423-60-5
EINECS: 215-834-2
UN: 1992
ADR: 3,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR