Tisk…

1-Bromo-2-butyne, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

427292-1G 427292-5G
SIAL427292-1GEA 1220 CZK
SIAL427292-1G SIAL427292-5G
1-Bromo-2-butyne, Sigma-Aldrich®
1-Bromo-2-butyne
Vzorec: C₄H₅Br
MW: 132,99 g/mol
MDL Number: MFCD00190233
CAS číslo: 3355-28-0
UN: 1993
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR