Tisk…

trans-3-Fluorocinnamic acid ≥98%

Dodavatel: Thermo Scientific
Varování

B20818.06 B20818.14 B20818.22
B20818.06EA 1100 CZK
B20818.06 B20818.14 B20818.22
trans-3-Fluorocinnamic acid ≥98%
trans-3-Fluorocinnamic acid

Purity: 98% CAS No.: 20595-30-6 Molecular Formula: C9H7FO2 Formula Weight: 166.15 Odour: Odourless Melting Point: 165-168° Beilstein Reference: 1365172 MDL No.: MFCD00004383

Vzorec: C₉H₇FO₂
MW: 166,15 g/mol
Bod tání: 165…168 °C
Storage Temperature: Ambient
MDL Number: MFCD00004383
CAS číslo: 20595-30-6
EINECS: 207-281-0
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR